Product Details

SNP ID
rs147827058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162373537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGTTGACTGGTTCTCATCAGC[C/T]GGATGATGGGTCTCTGATATTCTGA
Phenotype
MIM: 607030 MIM: 608169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCA PubMed Links

Gene Details

Gene
GCA
Gene Name
grancalcin
There are no transcripts associated with this gene.

Gene
KCNH7
Gene Name
potassium voltage-gated channel subfamily H member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033272.3 3400 Missense Mutation CAG,CGG Q1086R NP_150375.2
NM_173162.2 3400 Intron NP_775185.1
XM_011512109.2 3400 Missense Mutation CAG,CGG Q1094R XP_011510411.1
XM_017005218.1 3400 Missense Mutation CAG,CGG Q1091R XP_016860707.1
XM_017005219.1 3400 Missense Mutation CAG,CGG Q1083R XP_016860708.1
XM_017005220.1 3400 Missense Mutation CAG,CGG Q1079R XP_016860709.1
XM_017005221.1 3400 Intron XP_016860710.1

View Full Product Details