Product Details

SNP ID
rs147920517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:209433751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTCTTATCTAATGCTGTAAC[A/G]TAACTCAAACTGTGTCATTTTTTTT
Phenotype
MIM: 157130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP2 PubMed Links

Gene Details

Gene
MAP2
Gene Name
microtubule associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039538.1 Intron NP_001034627.1
NM_002374.3 Intron NP_002365.3
NM_031845.2 Intron NP_114033.2
NM_031847.2 Intron NP_114035.2
XM_005246555.1 Intron XP_005246612.1
XM_005246565.1 Intron XP_005246622.1
XM_005246566.1 Intron XP_005246623.1
XM_006712532.1 Intron XP_006712595.1
XM_011511186.1 Intron XP_011509488.1
XM_011511187.1 Intron XP_011509489.1
XM_011511188.1 Intron XP_011509490.1
XM_011511190.1 Intron XP_011509492.1
XM_011511191.1 Intron XP_011509493.1
XM_011511192.1 Intron XP_011509494.1
XM_011511193.1 Intron XP_011509495.1
XM_011511195.1 Intron XP_011509497.1
XM_011511196.2 Intron XP_011509498.1
XM_011511197.2 Intron XP_011509499.1
XM_017004111.1 Intron XP_016859600.1
XM_017004112.1 Intron XP_016859601.1
XM_017004113.1 Intron XP_016859602.1
XM_017004114.1 Intron XP_016859603.1
XM_017004115.1 Intron XP_016859604.1
XM_017004116.1 Intron XP_016859605.1
XM_017004117.1 Intron XP_016859606.1
XM_017004118.1 Intron XP_016859607.1
XM_017004119.1 Intron XP_016859608.1
XM_017004120.1 Intron XP_016859609.1
XM_017004121.1 Intron XP_016859610.1
XM_017004122.1 Intron XP_016859611.1
XM_017004123.1 Intron XP_016859612.1
XM_017004124.1 Intron XP_016859613.1
XM_017004125.1 Intron XP_016859614.1
XM_017004126.1 Intron XP_016859615.1
XM_017004127.1 Intron XP_016859616.1
XM_017004128.1 Intron XP_016859617.1
XM_017004129.1 Intron XP_016859618.1
XM_017004130.1 Intron XP_016859619.1
XM_017004131.1 Intron XP_016859620.1
XM_017004132.1 Intron XP_016859621.1
XM_017004133.1 Intron XP_016859622.1
XM_017004134.1 Intron XP_016859623.1
XM_017004135.1 Intron XP_016859624.1
XM_017004136.1 Intron XP_016859625.1
XM_017004137.1 Intron XP_016859626.1
XM_017004138.1 Intron XP_016859627.1
XM_017004139.1 Intron XP_016859628.1
XM_017004140.1 Intron XP_016859629.1

View Full Product Details