Product Details

SNP ID
rs148210519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11447603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATGCATGCTTAATTAAAATAC[C/T]GTCAGAATCACTGGTATATTTACCT
Phenotype
MIM: 602944
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
E2F6 PubMed Links

Gene Details

Gene
E2F6
Gene Name
E2F transcription factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278275.1 Intron NP_001265204.1
NM_001278276.1 Intron NP_001265205.1
NM_001278277.1 Intron NP_001265206.1
NM_001278278.1 Intron NP_001265207.1
NM_198256.3 Intron NP_937987.2
NM_212540.2 Intron NP_997705.1
XM_017003547.1 Intron XP_016859036.1
XM_017003548.1 Intron XP_016859037.1
XM_017003549.1 Intron XP_016859038.1

View Full Product Details