Product Details

SNP ID
rs148227562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73219014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCATGGCCCAGTCGTCTTTGTG[C/T]ATGGAAGTGAGCTGCATACATCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOTO PubMed Links

Gene Details

Gene
NOTO
Gene Name
notochord homeobox
There are no transcripts associated with this gene.

Gene
PRADC1
Gene Name
protease associated domain containing 1
There are no transcripts associated with this gene.

Gene
SMYD5
Gene Name
SMYD family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006062.2 Intron NP_006053.2
XM_006711918.3 Intron XP_006711981.1
XM_017003163.1 Intron XP_016858652.1

View Full Product Details