Product Details
- SNP ID
-
rs148320729
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:24017481 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGATGACTGTGGAGATGGCGGGA[A/T]CACTGATGGGGGCCACTGTCCACGG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MFSD2B
PubMed Links
Gene Details
- Gene
- MFSD2B
- Gene Name
- major facilitator superfamily domain containing 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080473.1 |
1742 |
Missense Mutation |
ACA,TCA |
T192S |
NP_001073942.1 |
XM_005264301.4 |
1742 |
Missense Mutation |
ACA,TCA |
T192S |
XP_005264358.1 |
XM_017004063.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859552.1 |
XM_017004064.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859553.1 |
XM_017004065.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859554.1 |
XM_017004066.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859555.1 |
XM_017004067.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859556.1 |
XM_017004068.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859557.1 |
XM_017004069.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859558.1 |
XM_017004070.1 |
1742 |
Missense Mutation |
ACA,TCA |
T223S |
XP_016859559.1 |
View Full Product Details