Product Details

SNP ID
rs148356487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45006352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCAGGGGGCGGCGGGCTGAGGA[G/T]CAGTGCGGGGCTGGATGATGAGTGG
Phenotype
MIM: 604994
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIX2 PubMed Links

Gene Details

Gene
SIX2
Gene Name
SIX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016932.4 987 Missense Mutation ATC,CTC I232L NP_058628.3
XM_005264100.3 987 Missense Mutation ATC,CTC I234L XP_005264157.1

View Full Product Details