Product Details
- SNP ID
-
rs148920749
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100393407 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCGGAGGGATAGTCGGGTATGA[C/T]ACCAGGTGGTCAATGCCAGCCTCTT
- Phenotype
-
MIM: 606376
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHST10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59148684] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHST10
- Gene Name
- carbohydrate sulfotransferase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004854.4 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
NP_004845.1 |
XM_011512207.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_011510509.1 |
XM_011512208.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_011510510.1 |
XM_011512210.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_011510512.1 |
XM_011512211.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_011510513.1 |
XM_011512212.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_011510514.1 |
XM_017005380.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_016860869.1 |
XM_017005381.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_016860870.1 |
XM_017005382.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_016860871.1 |
XM_017005383.1 |
1631 |
Silent Mutation |
GTA,GTG |
V303V |
XP_016860872.1 |
View Full Product Details