Product Details

SNP ID
rs149567782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216415000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTCAGGCAAAGGGAATATGGA[A/G]AAAGCCAGAAGAAATGCCCACAGCC
Phenotype
MIM: 606622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMARCAL1 PubMed Links

Gene Details

Gene
SMARCAL1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127207.1 440 Missense Mutation AAA,AGA K99R NP_001120679.1
NM_014140.3 440 Missense Mutation AAA,AGA K99R NP_054859.2
XM_005246631.2 440 Missense Mutation AAA,AGA K99R XP_005246688.1
XM_005246632.1 440 Missense Mutation AAA,AGA K99R XP_005246689.1
XM_017004228.1 440 UTR 5 XP_016859717.1

View Full Product Details