Product Details

SNP ID
rs149877713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24947731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGCACAGCAAGTTTCCGATAC[A/G]CTTTATTGACTTCATCCCTGGGAAA
Phenotype
MIM: 613527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC27 PubMed Links

Gene Details

Gene
DNAJC27
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198559.1 788 UTR 3 NP_001185488.1
NM_016544.2 788 Missense Mutation GCG,GTG A236V NP_057628.1
XM_011532902.1 788 Intron XP_011531204.1
XM_017004269.1 788 Missense Mutation GCG,GTG A165V XP_016859758.1
XM_017004270.1 788 Missense Mutation GCG,GTG A165V XP_016859759.1

View Full Product Details