Product Details
- SNP ID
-
rs150087096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:102187907 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTGCGGGTTGTCCATCGCCCTT[A/C]CACTGTCTGTCACAGCAGGTACGTT
- Phenotype
-
MIM: 147810
MIM: 604512
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL1R1
PubMed Links
Gene Details
- Gene
- IL1R1
- Gene Name
- interleukin 1 receptor type 1
There are no transcripts associated with this gene.
- Gene
- IL1RL2
- Gene Name
- interleukin 1 receptor like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003854.2 |
301 |
Missense Mutation |
ACA,CCA |
T14P |
NP_003845.2 |
XM_005264035.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_005264092.1 |
XM_005264036.1 |
301 |
Missense Mutation |
ACA,CCA |
T14P |
XP_005264093.1 |
XM_006712820.2 |
301 |
Missense Mutation |
TTA,TTC |
L91F |
XP_006712883.1 |
XM_006712822.2 |
301 |
Intron |
|
|
XP_006712885.1 |
XM_011512091.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_011510393.1 |
XM_011512092.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_011510394.1 |
XM_011512093.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_011510395.1 |
XM_011512094.1 |
301 |
Missense Mutation |
ACA,CCA |
T14P |
XP_011510396.1 |
XM_011512096.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_011510398.1 |
XM_017005174.1 |
301 |
Missense Mutation |
ACA,CCA |
T42P |
XP_016860663.1 |
XM_017005175.1 |
301 |
Missense Mutation |
TTA,TTC |
L91F |
XP_016860664.1 |
XM_017005176.1 |
301 |
Missense Mutation |
TTA,TTC |
L91F |
XP_016860665.1 |
XM_017005177.1 |
301 |
UTR 5 |
|
|
XP_016860666.1 |
XM_017005178.1 |
301 |
UTR 5 |
|
|
XP_016860667.1 |
XM_017005179.1 |
301 |
UTR 5 |
|
|
XP_016860668.1 |
XM_017005180.1 |
301 |
UTR 5 |
|
|
XP_016860669.1 |
View Full Product Details