Product Details
- SNP ID
-
rs150578544
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218164277 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGATCTTGAGGAATCCATGGCGA[A/G]AATTTTGGCCGATGAAGGCGTAGAT
- Phenotype
-
MIM: 146929
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CXCR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16858808] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CXCR1
- Gene Name
- C-X-C motif chemokine receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000634.2 |
1054 |
Missense Mutation |
TCT,TTT |
S312F |
NP_000625.1 |
View Full Product Details