Product Details

SNP ID
rs150617661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:121219023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGAGGGTACAAACATTTCCTA[A/C]CTGCAACCCTGGACCGGTAAGATGA
Phenotype
MIM: 609785
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TFCP2L1 PubMed Links

Gene Details

Gene
TFCP2L1
Gene Name
transcription factor CP2-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014553.2 6158 UTR 3 NP_055368.1
XM_017003902.1 6158 UTR 3 XP_016859391.1
XM_017003903.1 6158 UTR 3 XP_016859392.1
XM_017003904.1 6158 Intron XP_016859393.1
XM_017003905.1 6158 Intron XP_016859394.1

View Full Product Details