Product Details

SNP ID
rs151264953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27276757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACTGTGAAGGAAGCAGCACACC[A/G]GCTGTCCAAAAGTGAGATGAGCCTC
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 385 Missense Mutation CAG,CGG Q10R NP_001290056.1
NM_001303128.1 385 Missense Mutation CAG,CGG Q10R NP_001290057.1
NM_173650.2 385 Missense Mutation CAG,CGG Q10R NP_775921.1
XM_006711995.3 385 Missense Mutation CAG,CGG Q10R XP_006712058.1
XM_017003872.1 385 Missense Mutation CAG,CGG Q10R XP_016859361.1
XM_017003873.1 385 Missense Mutation CAG,CGG Q10R XP_016859362.1
XM_017003874.1 385 Missense Mutation CAG,CGG Q10R XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318949.1 385 Intron NP_001305878.1
NM_001318950.1 385 Intron NP_001305879.1
NM_001318951.1 385 Intron NP_001305880.1
NM_003459.4 385 Intron NP_003450.2
XM_005264547.1 385 Intron XP_005264604.1
XM_005264548.3 385 Intron XP_005264605.1
XM_006712100.2 385 Intron XP_006712163.1
XM_011533102.2 385 Intron XP_011531404.1
XM_011533103.2 385 Intron XP_011531405.1
XM_017004873.1 385 Intron XP_016860362.1
XM_017004874.1 385 Intron XP_016860363.1
XM_017004875.1 385 Intron XP_016860364.1
Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details