Product Details

SNP ID
rs151274887
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238849761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGGGTCCTTTTCACCAGACACC[C/T]TCCAAGTCCCTTTAGACAAGCTCGA
Phenotype
MIM: 607556
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC100287387 PubMed Links

Gene Details

Gene
LOC100287387
Gene Name
uncharacterized LOC100287387
There are no transcripts associated with this gene.

Gene
TWIST2
Gene Name
twist family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271893.3 Intron NP_001258822.1
NM_057179.2 Intron NP_476527.1

View Full Product Details