Product Details

SNP ID
rs137906016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32484022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTATTTTCAGGTTTTCTCAGCT[C/G]GGACTCCTGTCGTTTTTCATACAGC
Phenotype
MIM: 607889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CMTM6 PubMed Links

Gene Details

Gene
CMTM6
Gene Name
CKLF like MARVEL transmembrane domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017801.2 656 Missense Mutation CAG,GAG Q164E NP_060271.1

View Full Product Details