Product Details

SNP ID
rs138742466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24120359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTTCCTTTACTTGCGGCTGGCT[A/G]GATGGCTGTTTATATGCCATACTAT
Phenotype
MIM: 190160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 4333 UTR 3 NP_000452.2
NM_001128176.2 4333 UTR 3 NP_001121648.1
NM_001128177.1 4333 UTR 3 NP_001121649.1
NM_001252634.1 4333 UTR 3 NP_001239563.1
XM_005265421.4 4333 UTR 3 XP_005265478.1
XM_005265423.4 4333 UTR 3 XP_005265480.1
XM_005265424.3 4333 UTR 3 XP_005265481.1
XM_006713317.3 4333 UTR 3 XP_006713380.1
XM_006713318.3 4333 UTR 3 XP_006713381.1
XM_011534046.2 4333 UTR 3 XP_011532348.1
XM_011534047.2 4333 UTR 3 XP_011532349.1
XM_011534048.2 4333 UTR 3 XP_011532350.1
XM_011534049.2 4333 UTR 3 XP_011532351.1
XM_011534050.2 4333 UTR 3 XP_011532352.1
XM_011534051.2 4333 UTR 3 XP_011532353.1
XM_011534052.2 4333 UTR 3 XP_011532354.1
XM_011534053.2 4333 UTR 3 XP_011532355.1
XM_017007108.1 4333 UTR 3 XP_016862597.1
XM_017007109.1 4333 UTR 3 XP_016862598.1
XM_017007110.1 4333 UTR 3 XP_016862599.1
XM_017007111.1 4333 UTR 3 XP_016862600.1
XM_017007112.1 4333 UTR 3 XP_016862601.1
XM_017007113.1 4333 UTR 3 XP_016862602.1
XM_017007114.1 4333 UTR 3 XP_016862603.1
XM_017007115.1 4333 UTR 3 XP_016862604.1

View Full Product Details