Product Details

SNP ID
rs139461818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133928502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTGAGCAAGGTGGGGGCCCC[A/G]TGGGTGTGCTGGGTAGGGGTGGAGT
Phenotype
MIM: 601460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf36 PubMed Links

Gene Details

Gene
C3orf36
Gene Name
chromosome 3 open reading frame 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025041.2 1311 Missense Mutation ACG,ATG T101M NP_079317.2
Gene
SLCO2A1
Gene Name
solute carrier organic anion transporter family member 2A1
There are no transcripts associated with this gene.

View Full Product Details