Product Details

SNP ID
rs139527321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139233057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATATGGCTTTATTCACAGCTGAC[G/T]CACTTTGGGAGAGTCACGGCTTCTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PISRT1 PubMed Links

Gene Details

Gene
PISRT1
Gene Name
polled intersex syndrome regulated transcript 1 (non-protein coding RNA)
There are no transcripts associated with this gene.

View Full Product Details