Product Details

SNP ID
rs139543023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172889595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTTTGCTGAACAGTTTGAAGT[C/T]GCTTCATTTTTGTTTTTTGCCTTCG
Phenotype
MIM: 609856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA16 PubMed Links

Gene Details

Gene
SPATA16
Gene Name
spermatogenesis associated 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031955.5 1869 Missense Mutation NP_114161.3
XM_006713778.2 1869 Intron XP_006713841.1
XM_017007308.1 1869 Intron XP_016862797.1

View Full Product Details