Product Details

SNP ID
rs140127056
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38126203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGGATGGCATAGGCAGGTCCA[A/G]TGCCCATGATGTCAGGTGGGACCCC
Phenotype
MIM: 604054 MIM: 604050
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACAA1 PubMed Links

Gene Details

Gene
ACAA1
Gene Name
acetyl-CoA acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130410.1 1163 Intron NP_001123882.1
NM_001607.3 1163 Missense Mutation ACT,ATT T319I NP_001598.1
XM_006713122.1 1163 Intron XP_006713185.1
XM_006713123.2 1163 Missense Mutation ACT,ATT T167I XP_006713186.1
XM_011533650.2 1163 Missense Mutation ACT,ATT T174I XP_011531952.1
Gene
DLEC1
Gene Name
deleted in lung and esophageal cancer 1
There are no transcripts associated with this gene.

View Full Product Details