Product Details

SNP ID
rs140254923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53848667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTTTTTTCTCTCCCACAGACC[A/G]TTCAATGTGGCTCTGAAACTGGTAG
Phenotype
MIM: 605458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHDH PubMed Links

Gene Details

Gene
CHDH
Gene Name
choline dehydrogenase
There are no transcripts associated with this gene.

Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 100 Missense Mutation ATT,GTT I22V NP_061195.2
XM_005265310.4 100 Missense Mutation ATT,GTT I22V XP_005265367.1
XM_005265311.4 100 Missense Mutation ATT,GTT I22V XP_005265368.1
XM_005265312.4 100 Missense Mutation ATT,GTT I22V XP_005265369.1
XM_011533941.2 100 Missense Mutation ATT,GTT I22V XP_011532243.1
XM_017006804.1 100 UTR 5 XP_016862293.1
XM_017006805.1 100 UTR 5 XP_016862294.1
XM_017006806.1 100 UTR 5 XP_016862295.1
XM_017006807.1 100 Missense Mutation ATT,GTT I22V XP_016862296.1

View Full Product Details