Product Details

SNP ID
rs140579801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:99642538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCTGACTATGTAAAATATGAA[C/T]ACACACCTGACAATTCACCTTCGGT
Phenotype
MIM: 120251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL8A1 PubMed Links

Gene Details

Gene
COL8A1
Gene Name
collagen type VIII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001850.4 Intron NP_001841.2
NM_020351.3 Intron NP_065084.2
Gene
LOC105374008
Gene Name
uncharacterized LOC105374008
There are no transcripts associated with this gene.

View Full Product Details