Product Details

SNP ID
rs140875358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:72378496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTCTGGTTCTGCTGTTCTGA[C/T]CCTGCACTGGAGGTCACTGTGGATG
Phenotype
MIM: 607535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RYBP PubMed Links

Gene Details

Gene
RYBP
Gene Name
RING1 and YY1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012234.6 759 Silent Mutation GGA,GGG G192G NP_036366.3

View Full Product Details