Product Details

SNP ID
rs140924394
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187698690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCGCTGGGCTTCCAGTGAAC[C/G]ATGCCCTCCTGGCAGGCCTCACAGA
Phenotype
MIM: 609138
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100131635 PubMed Links

Gene Details

Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004312.2 1060 Missense Mutation ATC,ATG I162M NP_001004312.2
XM_017006301.1 1060 Missense Mutation ATC,ATG I162M XP_016861790.1
XM_017006302.1 1060 Missense Mutation ATC,ATG I162M XP_016861791.1

View Full Product Details