Product Details

SNP ID
rs141551640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44721150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGAGAAAAGCTTGCTTTTGAC[C/T]TCAAGCCTTGTTACACGTCTCAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF501 PubMed Links

Gene Details

Gene
ZNF501
Gene Name
zinc finger protein 501
There are no transcripts associated with this gene.

Gene
ZNF502
Gene Name
zinc finger protein 502
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134440.1 1830 Silent Mutation ACC,ACT T111T NP_001127912.1
NM_001134441.1 1830 Silent Mutation ACC,ACT T111T NP_001127913.1
NM_001134442.1 1830 Silent Mutation ACC,ACT T111T NP_001127914.1
NM_001282880.1 1830 Silent Mutation ACC,ACT T111T NP_001269809.1
NM_033210.4 1830 Silent Mutation ACC,ACT T111T NP_149987.2
XM_017007440.1 1830 Silent Mutation ACC,ACT T111T XP_016862929.1

View Full Product Details