Product Details

SNP ID
rs142736119
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151814251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTCCCAGATAACGTTGAGGAGC[C/T]ATGGAGAATGATGTGGATAAACGCA
Phenotype
MIM: 600338
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AADAC PubMed Links

Gene Details

Gene
AADAC
Gene Name
arylacetamide deacetylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001086.2 179 Missense Mutation CCA,CTA P30L NP_001077.2
XM_005247104.4 179 Missense Mutation CCA,CTA P30L XP_005247161.1
Gene
AADACL2-AS1
Gene Name
AADACL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details