Product Details
- SNP ID
-
rs143671992
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:187698925 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGACGCGCATGCGCACCGAGCCC[G/T]CCCGCTGGGCGCGGTCCAGGAACAT
- Phenotype
-
MIM: 609138
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100131635
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11707167] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100131635
- Gene Name
- hCG1645011-like
There are no transcripts associated with this gene.
- Gene
- RTP2
- Gene Name
- receptor transporter protein 2
View Full Product Details