Product Details

SNP ID
rs144461944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48466535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGGCAGGATTGTGCAGGGAAG[G/T]CCTGAGATGTGCTTCTGCCCACCCC
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271022.1 1016 Intron NP_001257951.1
NM_001271023.1 1016 Intron NP_001257952.1
NM_032166.3 1016 Intron NP_115542.2
NM_130384.2 1016 Intron NP_569055.1
Gene
SHISA5
Gene Name
shisa family member 5
There are no transcripts associated with this gene.

Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 1016 Intron NP_009179.2
NM_016381.5 1016 Missense Mutation AGG,AGT R15S NP_057465.1
NM_033629.4 1016 Intron NP_338599.1

View Full Product Details