Product Details

SNP ID
rs145306147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:157261245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCATCCTTGGCCTTAAAGAC[A/G]TAGGTTTTATTGTCTGTGAAGATTT
Phenotype
MIM: 609594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VEPH1 PubMed Links

Gene Details

Gene
VEPH1
Gene Name
ventricular zone expressed PH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167911.1 2449 Silent Mutation TAC,TAT Y752Y NP_001161383.1
NM_001167912.1 2449 Silent Mutation TAC,TAT Y797Y NP_001161384.1
NM_001167915.1 2449 Intron NP_001161387.1
NM_001167916.1 2449 Intron NP_001161388.1
NM_001167917.1 2449 Intron NP_001161389.1
NM_024621.2 2449 Silent Mutation TAC,TAT Y797Y NP_078897.2
XM_011513134.2 2449 Silent Mutation TAC,TAT Y808Y XP_011511436.1
XM_011513135.1 2449 Silent Mutation TAC,TAT Y752Y XP_011511437.1

View Full Product Details