Product Details

SNP ID
rs145434330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48466770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACGGAGCTGTGCCTGCTGGCT[C/G]TCCACAGATGTGCCCTGGAGAGCCC
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
There are no transcripts associated with this gene.

Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 1251 Missense Mutation CTC,GTC L29V NP_009179.2
NM_016381.5 1251 Missense Mutation CTC,GTC L94V NP_057465.1
NM_033629.4 1251 Missense Mutation CTC,GTC L39V NP_338599.1

View Full Product Details