Product Details

SNP ID
rs145442490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48165678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTATTCATATGGGTATCGACAGTC[A/G]ATGATAACAAACTCTTTAATGAGGT
Phenotype
MIM: 116947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC25A PubMed Links

Gene Details

Gene
CDC25A
Gene Name
cell division cycle 25A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001789.2 1605 Silent Mutation ATC,ATT I383I NP_001780.2
NM_201567.1 1605 Silent Mutation ATC,ATT I343I NP_963861.1
XM_006713434.2 1605 Silent Mutation ATC,ATT I382I XP_006713497.1
XM_006713435.2 1605 Silent Mutation ATC,ATT I342I XP_006713498.1
XM_011534316.1 1605 Silent Mutation ATC,ATT I383I XP_011532618.1

View Full Product Details