Product Details

SNP ID
rs145951517
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111675255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTAACAACAGGGATGCTAGCA[G/T]TTAGGAAGGCCAGGAGGAAACTCAG
Phenotype
MIM: 606037 MIM: 617015
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CD96 PubMed Links

Gene Details

Gene
CD96
Gene Name
CD96 molecule
There are no transcripts associated with this gene.

Gene
PLCXD2
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185106.1 580 Missense Mutation GTT,TTT V4F NP_001172035.1
NM_153268.3 580 Missense Mutation GTT,TTT V4F NP_695000.1
Gene
PLCXD2-AS1
Gene Name
PLCXD2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details