Product Details

SNP ID
rs146072078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120750703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAAAGAGGAGGATATGCTGGAG[C/G]TGCTCAAGTTTGATCGGAAGCAACT
Phenotype
MIM: 189962
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF2E1 PubMed Links

Gene Details

Gene
GTF2E1
Gene Name
general transcription factor IIE subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005513.2 336 Missense Mutation CTG,GTG L51V NP_005504.2
XM_011512744.2 336 Missense Mutation CTG,GTG L51V XP_011511046.1
XM_011512745.2 336 Missense Mutation CTG,GTG L51V XP_011511047.1
Gene
RABL3
Gene Name
RAB, member of RAS oncogene family like 3
There are no transcripts associated with this gene.

View Full Product Details