Product Details

SNP ID
rs146298087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138348064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCAACGAAGTTTGGGCCCTG[G/T]AACGGTGCACCCAGAAGGCCTGCGG
Phenotype
MIM: 608435
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRAS PubMed Links

Gene Details

Gene
MRAS
Gene Name
muscle RAS oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085049.2 417 Intron NP_001078518.1
NM_001252090.1 417 Intron NP_001239019.1
NM_001252091.1 417 UTR 5 NP_001239020.1
NM_001252092.1 417 Intron NP_001239021.1
NM_001252093.1 417 Intron NP_001239022.1
NM_012219.4 417 UTR 5 NP_036351.3
XM_005247228.1 417 Intron XP_005247285.1
XM_005247229.2 417 Intron XP_005247286.1
XM_017005887.1 417 Intron XP_016861376.1

View Full Product Details