Product Details

SNP ID
rs146327020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48164369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAACAATGACACGCTTGCCATC[A/C]GTAGGTACAATGGGCTTCTTCAATA
Phenotype
MIM: 116947
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC25A PubMed Links

Gene Details

Gene
CDC25A
Gene Name
cell division cycle 25A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001789.2 1716 Silent Mutation ACG,ACT T420T NP_001780.2
NM_201567.1 1716 Silent Mutation ACG,ACT T380T NP_963861.1
XM_006713434.2 1716 Silent Mutation ACG,ACT T419T XP_006713497.1
XM_006713435.2 1716 Silent Mutation ACG,ACT T379T XP_006713498.1
XM_011534316.1 1716 Silent Mutation ACG,ACT T420T XP_011532618.1

View Full Product Details