Product Details

SNP ID
rs146586055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9712824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTCTACAGGAGCAATGAGGAT[G/T]GCACGTGAGTCACTGCCATCAGGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CPNE9 PubMed Links

Gene Details

Gene
CPNE9
Gene Name
copine family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308388.1 888 Missense Mutation GGC,TGC G181C NP_001295317.1
NM_153635.2 888 Missense Mutation GGC,TGC G181C NP_705899.2
XM_011533386.2 888 Missense Mutation GGC,TGC G290C XP_011531688.1
XM_011533388.2 888 Missense Mutation GGC,TGC G290C XP_011531690.1
XM_011533389.2 888 Missense Mutation GGC,TGC G290C XP_011531691.1
XM_017005747.1 888 UTR 5 XP_016861236.1

View Full Product Details