Product Details
- SNP ID
-
rs146640916
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:195866984 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCACTTTGTGGCACTCCCCTCT[A/G]GGCCGCAGACCCAGCCCGAAGAGCT
- Phenotype
-
MIM: 606994
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929697
PubMed Links
Gene Details
- Gene
- LOC101929697
- Gene Name
- uncharacterized LOC101929697
There are no transcripts associated with this gene.
- Gene
- TNK2
- Gene Name
- tyrosine kinase non receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010938.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1053P |
NP_001010938.1 |
NM_001308046.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1007P |
NP_001294975.1 |
NM_005781.4 |
3558 |
Silent Mutation |
CCC,CCT |
P1005P |
NP_005772.3 |
XM_005269268.3 |
3558 |
Silent Mutation |
CCC,CCT |
P1085P |
XP_005269325.1 |
XM_005269270.3 |
3558 |
Silent Mutation |
CCC,CCT |
P1022P |
XP_005269327.1 |
XM_005269274.3 |
3558 |
Silent Mutation |
CCC,CCT |
P783P |
XP_005269331.1 |
XM_005269275.3 |
3558 |
Silent Mutation |
CCC,CCT |
P708P |
XP_005269332.1 |
XM_011512317.2 |
3558 |
Silent Mutation |
CCC,CCT |
P1186P |
XP_011510619.1 |
XM_011512318.2 |
3558 |
Silent Mutation |
CCC,CCT |
P1052P |
XP_011510620.2 |
XM_011512320.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1022P |
XP_011510622.1 |
XM_011512321.2 |
3558 |
Silent Mutation |
CCC,CCT |
P946P |
XP_011510623.1 |
XM_017005508.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1039P |
XP_016860997.1 |
XM_017005509.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1037P |
XP_016860998.1 |
XM_017005510.1 |
3558 |
Silent Mutation |
CCC,CCT |
P1022P |
XP_016860999.1 |
View Full Product Details