Product Details

SNP ID
rs146953866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184178178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCTATCACTCTCCTCTTCTTGC[C/T]GGCGTCATTTCTCTCATCCCATCTC
Phenotype
MIM: 601024 MIM: 601368
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCF3 PubMed Links

Gene Details

Gene
ABCF3
Gene Name
ATP binding cassette subfamily F member 3
There are no transcripts associated with this gene.

Gene
AP2M1
Gene Name
adaptor related protein complex 2 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025205.1 Intron NP_001020376.1
NM_001311198.1 Intron NP_001298127.1
NM_004068.3 Intron NP_004059.2
Gene
DVL3
Gene Name
dishevelled segment polarity protein 3
There are no transcripts associated with this gene.

View Full Product Details