Product Details
- SNP ID
-
rs147002334
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:127675975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGCCCTGCCTCCCCCAGGAGCC[A/C]GCTGCGAGGCCAACACCTTCGATGG
- Phenotype
-
MIM: 608308
MIM: 616627
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABTB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35703629] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABTB1
- Gene Name
- ankyrin repeat and BTB domain containing 1
- Gene
- PODXL2
- Gene Name
- podocalyxin like 2
There are no transcripts associated with this gene.
View Full Product Details