Product Details

SNP ID
rs147434424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150926909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGATGGGTAATTCAGGGGAAAAA[A/C]AAAGTTGACCTGGGTCATGCTTGGT
Phenotype
MIM: 606397
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLRN1 PubMed Links

Gene Details

Gene
CLRN1
Gene Name
clarin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195794.1 2056 UTR 3 NP_001182723.1
NM_001256819.1 2056 UTR 3 NP_001243748.1
NM_052995.2 2056 Intron NP_443721.1
NM_174878.2 2056 UTR 3 NP_777367.1

View Full Product Details