Product Details

SNP ID
rs147479799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:154276185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAAGCTGTCAGCTGTAATATC[C/T]ATCCTGGAATCGTGGCGGAAAGTTC
Phenotype
MIM: 612767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHX36 PubMed Links

Gene Details

Gene
DHX36
Gene Name
DEAH-box helicase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114397.1 3094 Missense Mutation AGA,GGA R991G NP_001107869.1
NM_020865.2 3094 Missense Mutation AGA,GGA R1005G NP_065916.2

View Full Product Details