Product Details
- SNP ID
-
rs147633710
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:113965214 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGTATCTGATTGCTGATATAGCT[C/T]GTGGTACCTTCCAGGTACCTGGAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC191
PubMed Links
Gene Details
- Gene
- CCDC191
- Gene Name
- coiled-coil domain containing 191
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020817.1 |
2841 |
Missense Mutation |
AAG,GAG |
K918E |
NP_065868.1 |
XM_005247681.3 |
2841 |
Missense Mutation |
AAG,GAG |
K597E |
XP_005247738.1 |
XM_006713715.1 |
2841 |
Missense Mutation |
AAG,GAG |
K905E |
XP_006713778.1 |
XM_011513051.1 |
2841 |
Missense Mutation |
AAG,GAG |
K876E |
XP_011511353.1 |
XM_011513052.2 |
2841 |
Missense Mutation |
AAG,GAG |
K856E |
XP_011511354.1 |
XM_011513053.1 |
2841 |
Missense Mutation |
AAG,GAG |
K833E |
XP_011511355.1 |
XM_011513054.2 |
2841 |
Missense Mutation |
AAG,GAG |
K833E |
XP_011511356.1 |
XM_017006956.1 |
2841 |
Missense Mutation |
AAG,GAG |
K896E |
XP_016862445.1 |
XM_017006957.1 |
2841 |
Missense Mutation |
AAG,GAG |
K833E |
XP_016862446.1 |
XM_017006958.1 |
2841 |
Missense Mutation |
AAG,GAG |
K833E |
XP_016862447.1 |
XM_017006959.1 |
2841 |
Missense Mutation |
AAG,GAG |
K518E |
XP_016862448.1 |
- Gene
- ZDHHC23
- Gene Name
- zinc finger DHHC-type containing 23
View Full Product Details