Product Details

SNP ID
rs148464732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131383254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAATTCCTTTATTGGCTCTGC[A/G]CGGGAGCAGTTACTTGAAGCTCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC339874 PubMed Links

Gene Details

Gene
LOC339874
Gene Name
uncharacterized LOC339874
There are no transcripts associated with this gene.

Gene
NUDT16
Gene Name
nudix hydrolase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171905.1 438 Silent Mutation GCA,GCG A121A NP_001165376.1
NM_001171906.1 438 Intron NP_001165377.1
NM_152395.2 438 Silent Mutation GCA,GCG A167A NP_689608.2

View Full Product Details