Product Details

SNP ID
rs149053779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128059976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCGTTCTCAGCCGAGCCCCTTG[A/G]AGGTAGTATCACTGCTCTTCATCTT
Phenotype
MIM: 603449 MIM: 609213
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RUVBL1 PubMed Links

Gene Details

Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
There are no transcripts associated with this gene.

Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 Intron NP_037468.1

View Full Product Details