Product Details
- SNP ID
-
rs149927939
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:127604748 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCAGCGTGACCGGGAGGCTGGCC[A/G]GGGCCTGGGCCGCATGCGCCGTGGG
- Phenotype
-
MIM: 116945
MIM: 608336
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MCM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2257836] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MCM2
- Gene Name
- minichromosome maintenance complex component 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004526.3 |
487 |
Missense Mutation |
CAG,CGG |
Q126R |
NP_004517.2 |
- Gene
- TPRA1
- Gene Name
- transmembrane protein adipocyte associated 1
There are no transcripts associated with this gene.
View Full Product Details