Product Details
- SNP ID
-
rs150366825
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:127573674 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCAGCTGAGGCCCCAGCAGCCCC[C/T]GCAGCCTCCAGGCCCTCCCGCCGGG
- Phenotype
-
MIM: 608336
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6825
PubMed Links
Gene Details
- Gene
- MIR6825
- Gene Name
- microRNA 6825
There are no transcripts associated with this gene.
- Gene
- TPRA1
- Gene Name
- transmembrane protein adipocyte associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136053.2 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
NP_001129525.1 |
NM_001142646.2 |
1370 |
Missense Mutation |
AGG,GGG |
R269G |
NP_001136118.1 |
XM_006713495.3 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_006713558.1 |
XM_006713496.2 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_006713559.1 |
XM_006713497.3 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_006713560.1 |
XM_011512422.1 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_011510724.1 |
XM_017005707.1 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_016861196.1 |
XM_017005708.1 |
1370 |
Silent Mutation |
GCA,GCG |
A323A |
XP_016861197.1 |
XM_017005709.1 |
1370 |
Missense Mutation |
AGG,GGG |
R327G |
XP_016861198.1 |
XM_017005710.1 |
1370 |
Silent Mutation |
GCA,GCG |
A286A |
XP_016861199.1 |
View Full Product Details