Product Details

SNP ID
rs150767504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125230621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTCTTTATGAGAACAGTCTT[C/G]GGATAACATTAAATAAAAGAAAAAA
Phenotype
MIM: 601897
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF148 PubMed Links

Gene Details

Gene
ZNF148
Gene Name
zinc finger protein 148
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021964.2 4591 UTR 3 NP_068799.2

View Full Product Details