Product Details
- SNP ID
-
rs151014440
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:47414198 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTACCTGCTGAATGGAGTAGAA[C/T]TTGATGCCTGTGCTGCGGTCCCAGA
- Phenotype
-
MIM: 606584
MIM: 601510
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PTPN23
PubMed Links
Gene Details
- Gene
- PTPN23
- Gene Name
- protein tyrosine phosphatase, non-receptor type 23
- Gene
- SCAP
- Gene Name
- SREBF chaperone
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320044.1 |
3504 |
Silent Mutation |
AAA,AAG |
K936K |
NP_001306973.1 |
NM_012235.3 |
3504 |
Silent Mutation |
AAA,AAG |
K1192K |
NP_036367.2 |
XM_005264967.1 |
3504 |
Silent Mutation |
AAA,AAG |
K1191K |
XP_005265024.1 |
XM_005264968.1 |
3504 |
Silent Mutation |
AAA,AAG |
K937K |
XP_005265025.1 |
XM_005264971.1 |
3504 |
Silent Mutation |
AAA,AAG |
K800K |
XP_005265028.1 |
XM_011533501.1 |
3504 |
Silent Mutation |
AAA,AAG |
K1192K |
XP_011531803.1 |
XM_011533502.2 |
3504 |
Silent Mutation |
AAA,AAG |
K811K |
XP_011531804.1 |
XM_017005918.1 |
3504 |
Silent Mutation |
AAA,AAG |
K1192K |
XP_016861407.1 |
XM_017005919.1 |
3504 |
Silent Mutation |
AAA,AAG |
K962K |
XP_016861408.1 |
XM_017005920.1 |
3504 |
Silent Mutation |
AAA,AAG |
K961K |
XP_016861409.1 |
XM_017005921.1 |
3504 |
Silent Mutation |
AAA,AAG |
K799K |
XP_016861410.1 |
View Full Product Details