Product Details

SNP ID
rs151051255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49830338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCACTCAAGGCCAAATTCCAA[C/T]GCTCCTCAAGTCCTCGGGCACCTCA
Phenotype
MIM: 605958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAIP PubMed Links

Gene Details

Gene
TRAIP
Gene Name
TRAF interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005879.2 Intron NP_005870.2
XM_011533264.1 Intron XP_011531566.1
XM_017005526.1 Intron XP_016861015.1

View Full Product Details