Product Details

SNP ID
rs151222302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169084452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTAAATTACATAAAACTTTAA[C/T]GAAGAAAAAAATTAAATCTGTAATT
Phenotype
MIM: 165215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MECOM PubMed Links

Gene Details

Gene
MECOM
Gene Name
MDS1 and EVI1 complex locus
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105077.3 3878 UTR 3 NP_001098547.3
NM_001105078.3 3878 UTR 3 NP_001098548.2
NM_001163999.1 3878 UTR 3 NP_001157471.1
NM_001164000.1 3878 UTR 3 NP_001157472.1
NM_001205194.1 3878 UTR 3 NP_001192123.1
NM_004991.3 3878 UTR 3 NP_004982.2
NM_005241.3 3878 UTR 3 NP_005232.2
XM_005247213.3 3878 UTR 3 XP_005247270.1
XM_005247214.3 3878 UTR 3 XP_005247271.1
XM_005247215.3 3878 UTR 3 XP_005247272.1
XM_005247219.2 3878 UTR 3 XP_005247276.1
XM_005247220.2 3878 UTR 3 XP_005247277.1
XM_005247221.2 3878 UTR 3 XP_005247278.1
XM_005247223.2 3878 UTR 3 XP_005247280.1
XM_005247224.3 3878 UTR 3 XP_005247281.1
XM_005247225.3 3878 UTR 3 XP_005247282.1
XM_005247226.3 3878 UTR 3 XP_005247283.1
XM_011512546.2 3878 UTR 3 XP_011510848.1
XM_017005874.1 3878 UTR 3 XP_016861363.1
XM_017005875.1 3878 UTR 3 XP_016861364.1
XM_017005876.1 3878 UTR 3 XP_016861365.1
XM_017005877.1 3878 UTR 3 XP_016861366.1
XM_017005878.1 3878 UTR 3 XP_016861367.1

View Full Product Details